VWR®, Refractive index standards
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
85133.015EA 88.3 EUR
85133.015 85134.015 85135.015 85136.015 85137.015 85138.015 85114.015 85115.015 85116.015 85117.015 85118.015 85119.015 85120.015 85121.015 85122.015 85123.015 85124.015 85125.015 85126.015 85127.015 85128.015 85129.015 85130.015 85131.015 85132.015
VWR®, Refractive index standards
Standaarden Standaarden voor brekingsindexbepaling
Refractive index of each standard is verified using a calibrated and temperature controlled refractometer.
  • Standards are prepared gravimetrically on weight/weight basis
  • Solute (sucrose) and solvent (water) are weighed on a calibrated balance using OIML traceable weights
Certificaten: Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.