Ammoniumcarbonaat aqueous solution USP Test Solution (TS)
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Salt of hartshorn
85328.180EA 64.4 EUR
85328.180 85328.260
Ammoniumcarbonaat aqueous solution USP Test Solution (TS)
Ammonium carbonate
Ready to use test solution (TS).
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Manufactured with controlled processes
  • Independent, traceable and certified
  • Certificates of analysis and safety data sheets available
Formule: (NH₄)₂CO₃
MW: 96.09 g/mol
Smeltpunt: 58
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010890
CAS nummer: 506-87-6
EINECS: 0
Merck Index: 13,00508