Substrate buffers, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34064
34062
PIER34064EA
98.75
EUR
PIER34064
PIER34062
Substrate buffers, Pierce™
Buffers
Enzymbuffers
These stable peroxide and diethanolamine substrate buffers produce optimised, reliable and convenient stock solutions for HRP and alkaline phosphatase assays detection respectively.
- Ready to use
- Minimises assay-to-assay variability