Pierce™ OPD (o-phenylenediamine Dihydrochloride)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34006 34005
PIER34006EA 214 EUR
PIER34006 PIER34005
Pierce™ OPD (o-phenylenediamine Dihydrochloride)
Reagentia voor elektroforese Western Blotting Reagents
OPD (o-phenylenediamine dihydrochloride) is a water-soluble substrate for horseradish peroxidase (HRP) that produces a yellow-orange product detectable at 492 nm by ELISA plate readers.
Order Now
Specificaties
Environmentally Preferable