VWR®, Density standards referenced at 40 °C
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
85237.180EA 107 EUR
85237.180 85241.180 85243.180 85244.180 85245.180 85248.180 85250.180 85252.180
VWR®, Density standards referenced at 40 °C
Standaarden Standaarden voor dichtheidsbepaling
These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.
  • High accuracy across an extensive density range (0,6752 - 3,0852 g/ml) at 40 °C
  • No toxic heavy metals used in any formulation
  • Can be used with any brand or type of density measuring instrument
  • Supplied in a high quality, tamper-proof, amber glass bottle
These standards are prepared gravimetrically (solute and solvent) using OIML traceable weights. The density values of the standards are established using fully calibrated reference pycnometers.
Certificaten: In accordance with ASTM D1480-12 for testing of density or relative density. A certificate of calibration is issued in accordance with the requirements of ISO/IEC 17025.