Potassium carbonate in aqueous solution USP Test Solution (TS)
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Potash
85365.180EA 27.5 EUR
85365.180 85365.260
Potassium carbonate in aqueous solution USP Test Solution (TS)
Potassium carbonate
Ready to use test solution (TS).
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Formule: K₂CO₃
MW: 138.21 g/mol
Smeltpunt: 891
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011382
CAS nummer: 584-08-7
EINECS: 0
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM