Potassium iodide 16,5% w/v aqueous solution USP Test Solution (TS)
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Dipotassium diiodide, Kali iodide
85366.180EA 146 EUR
85366.180 85366.260
Potassium iodide 16,5% w/v aqueous solution USP Test Solution (TS)
Potassium iodide
Ready to use test solution (TS).
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Formule: Kl
MW: 166 g/mol
Kookpunt: 1330
Smeltpunt: 686
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011405
CAS nummer: 7681-11-0
EINECS: 0
Merck Index: 13,07727