VWR® Acetate Buffer USP Test Solution (TS)
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
85323.180EA
129
EUR
85323.180
85323.260
VWR® Acetate Buffer USP Test Solution (TS)
Buffers
pH-buffers
Ready to use test solutions (TS).
- Produced according to Pharmacopoeia requirements
- Reduces time and expenses
- Independent, traceable and certified
- Conforms to USP 34
- Certificates of analysis and safety data sheets available