Test kits, photometric, ammonium, LCK cuvette tests
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LCK303 LCK304 LCK305 LCK302 LCK502 LCK503 LCK505 LCK504
705-3006EA 155 EUR
705-3006 705-3007 705-3008 705-3005 HACHLCK502 HACHLCK503 HACHLCK505 HACHLCK504
Test kits, photometric, ammonium, LCK cuvette tests
Milieutestkits
Cuvette tests for water testing applications.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Shelf life: 18 months from production date.
Storage conditions: 2 to 8 °C (keep refrigerated)
Certificaten: ISO 7150-1, DIN 38406 E5-1
Informatie over toebehoren: Cuvette tests are officially approved for many legally required consent limits, with the help of Addista standard solutions and round-robin test solutions, they provide the assurance you need.