DNA plates for genomic/plasmid DNA extraction, E-Z® 96
Leverancier: OMEGA BIO-TEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BD96-01 BD96-02
IBD96-01EA 337 EUR
IBD96-01 IBD96-02
DNA plates for genomic/plasmid DNA extraction, E-Z® 96
Reagentia voor nucleïnezuren Nucleic Acid Purification Kits and Reagents
E-Z® 96 DNA plates are silica glass fibre plates can bind up to 50 μg of genomic DNA or 20 μg of plasmid DNA per prep. Each well has a capacity of 1 ml and the plates are semi skirted. E-Z 96 DNA plates can be processed in vacuum manifolds and/or by centrifugation.