DNA mini columns for genomic/plasmid DNA extraction, Hi-Bind®
Leverancier: OMEGA BIO-TEK
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DNACOL-01
DNACOL-02
DNACOL-01EA
48.9
EUR
DNACOL-01
DNACOL-02
DNA mini columns for genomic/plasmid DNA extraction, Hi-Bind®
Reagentia voor nucleïnezuren
Nucleic Acid Purification Kits and Reagents
Hi-Bind® DNA mini columns are silica glass fibre columns that can bind up to 100 μg of genomic DNA or 35 μg of plasmid DNA per prep. Columns feature an attached lid and a vacuum luer for processing with vacuum manifolds and by centrifugation.