Anti-IgG2b Goat Polyclonal Antibody (CF™ 405S)
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20382-250UL 20382-50UL
BTIU20382-250ULEA 385 EUR
BTIU20382-250UL BTIU20382-50UL
Anti-IgG2b Goat Polyclonal Antibody (CF™ 405S)
Antilichamen
Goat anti-mouse IgG2b is an affinity-purified antibody that reacts with Fc portion of the heavy chain of mouse IgG2b. To minimize cross reactivity, the antibody is cross-adsorbed against other mouse IgG subclasses (IgG1, IgG2a, IgG3), and human, bovine and rabbit serum proteins. CF™405S is a blue fluorescent dye with an absorption peak wavelength that nearly coincides with the 405 nm blue diode laser line. In addition, the emission peak wavelength of the dye well centers within the blue detection window of BD flow cytometers. As a result, flow cytometry analysis using CF™405S results in much brighter signal compared to Alexa Fluor® 405.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.
Type: Secondary
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: CF405S
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse