Anti-IgG Goat Polyclonal Antibody (CF™ 770)

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

20383-50UL 20383-250UL
BTIU20383-50ULEA 151 EUR
BTIU20383-50UL BTIU20383-250UL
Anti-IgG Goat Polyclonal Antibody (CF™ 770)
Antilichamen
This product is prepared by labeling highly cross-adsorbed goat anti-rat IgG (H+L) with the dye CF™770. CF™770 is one of our three outstanding near-IR CF™ dyes developed by Biotium. Near-IR CF™ dyes are superior to other similar near-IR dyes, such as IRDye® 800CW and the DyLight® 800. Near-IR CF™ dyes have combined advantages in brightness, photostability, specificity and novel features ideal for in vivo imaging, near-IR Western blotting and flow cytometry. To minimize crossreactivity, the antibody has been adsorbed against human, bovine, horse, and rabbit serum proteins.

DyLight® is a registered trademark of Thermo Fisher Scientific. IRDye® is a registered trademark of LI-COR® Bioscience.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF770
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR