Reverse osmosis water purification system, Barnstead™ Pacific™ RO
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
50132388 50132386 50132387
BARN50132388EA 8140 EUR
BARN50132388 BARN50132386 BARN50132387
Reverse osmosis water purification system, Barnstead™ Pacific™ RO
Waterzuiveringssystemen Puur- en ultrapuur-watersystemen
This system is a modularly designed laboratory reverse osmosis system specifically developed for functional and economical production of high purity water. Ideal for routine laboratory work: Rinsing lab glassware, supplying autoclaves, water baths and incubators, preparing and diluting buffers and reagents and general biotechnology.
  • Space-saving design allows system to sit on the laboratory bench or be mounted on the wall
  • Microprocessor control offers automatic operation with continual monitoring of all important parameters
  • Automatic return to operating
More Product Information
Order Now
Specificaties
Model Pacific™ RO 20 Pacific™ RO 7 Pacific™ RO 12
Environmentally Preferable
Debiet 20 L/h 7 L/h 12 L/h
Bacteriën 99% 99% 99%
b×d×h 372×330×603 mm 372×330×603 mm 372×330×603 mm
Gewicht 21 kg 21 kg 21 kg