Transfer pipettes, narrow stem, Samco™
Leverancier: SAMCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
241 242 251-1S 251-20S 251 252
SAMC241EA 0 EUR
SAMC241 SAMC242 SAMC251-1S SAMC251-20S SAMC251 SAMC252
Transfer pipettes, narrow stem, Samco™
Pipetten Pasteur-/transferpipetten
These pipettes made of shatterproof, non toxic PE prevent cross-contamination and are used for blood bank applications such as sedimentation procedures.
  • Low-affinity surface reduces loss of cells and valuable proteins due to binding
  • Can be frozen in liquid nitrogen, or heat sealed for storage
  • No need for plugging, packaging and autoclaving
The single-unit design features a built-in pipette bulb, and a flexible stem bends to sample from narrow or small tubes.