All-day cooling and freezing workstations, Corning® CoolBox™ XT
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
432021 432022 432023 432024 432025 432026 432027 432028
479-1863EA 594.2 EUR
479-1863 479-1864 479-1865 479-1866 479-1921 479-1922 479-1923 479-1924
All-day cooling and freezing workstations, Corning® CoolBox™ XT
Koelers Tafelmodel koelers
Ideaal voor het overbrengen van monsters van het ene labo naar het andere, of in het veld.
  • Over 16 hours of 0,5 to 4 °C cooling with lid closed and over 10 hours with the lid open
  • Reduces contamination by eliminating ice
  • Easy to clean with lab detergents or bleach
  • Patent-pending dual phase conductive XT Cooling core ensures stable and consistent sample cooling
  • Portable XT CoolBox systems can be used when a brief transfer of temperature sensitive samples to a robotic device is needed
  • May be used with dry ice in place of the Cooling Core to create a portable snap-freezing workstation
CoolBox™ XT ice-free bench top systems provide sample cooling or freezing without ice, electricity or batteries.
CoolBox XT workstations cool via thermoconductivity. Place the CoolRack module on top of the pre-frozen cooling core, included in the CoolBox XT and the samples achieve 0 °C in less than three minutes.
Ontwerp uw eigen CoolBox systeem. CoolBox XT bevat één koelkern en is geschikt voor één monstermodule. CoolBox 2XT bevat twee koelkernen en is geschikt voor twee monstermodules. Bestel CoolRack modules afzonderlijk gebaseerd op de vereiste buisjes- of plaatgrootte.
Bestelinformatie: See accessories for additional cooling cores as well as an optional freezing core for sample processing below 0 °C. Purchase CoolRack sample modules separately.