Bottles, wide neck, amber, low particle
Leverancier: EP SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
130-005/LP 122-16A/LP 121-08A/LP
EPSC130-005/LPEA 697 EUR
EPSC130-005/LP EPSC122-16A/LP EPSC121-08A/LP
Bottles, wide neck, amber, low particle
Flessen Glazen flessen
Glass, with PP screw cap.
  • Amber glass protects light-sensitive chemicals
  • PP caps contain no adhesives
  • Assembled caps
Protect the quality of your products or lab samples. Particle-certified glass containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter >0,5 µm. Ideal for preparation and storage of standards, off-line D.I. water and chemical sampling for particle analysis, and storage and packaging of chemicals, reagents, and powders.