Atropine sulphate monohydrate 97+%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
226680100 226680250 226681000
ACRO226680100EA 46.1 EUR
ACRO226680100 ACRO226680250 ACRO226681000
Atropine sulphate monohydrate 97+%
Atropine sulfate monohydrate
CAS No.: 5908-99-6
Formule: C₁₇H₂₃NO₃
MW: 676.82 g/mol
Smeltpunt: >187 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00074815
CAS nummer: 5908-99-6
UN: 1544
ADR: 6.1 II
Order Now
Specification Test Results
Appearance White Powder or crystals
Infrared spectrum Conforms
Water 2.0 to 4.0 % (K.F.)
Residue after ignition ≤0.2 %
Specific optical rotation -0.50° to +0.05° (25°C, 589 nm) (c=10, water) (on anhydrous basis)
Titration with HClO4 97.0 to 103.0 % (On anhydrous basis)