3,3',5,5'-Tetramethylbenzidine (TMB) Easy, liquid substrate
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
328030300 328031000
ACRO328030300EA 44 EUR
ACRO328030300 ACRO328031000
3,3',5,5'-Tetramethylbenzidine (TMB) Easy, liquid substrate
3,3',5,5'-Tetramethylbenzidine (TMB)
Formule: C₁₆H₂₀N₂
MW: 240.35 g/mol
Kookpunt: 402
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007748
CAS nummer: 54827-17-7
Order Now
Specification Test Results
Appearance Clear colorless to yellow-green to light blue Solution
Absorbance (Background level)