Iron(III) sulphate pentahydrate 97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
ferric sulphate hexahydrate, ferric sulphate, ferric sulphate heptahydrate
345230050
345235000
345230051
345231000
ACRO345230050EA
48.6
EUR
ACRO345230050
ACRO345235000
ACRO345230051
ACRO345231000
Iron(III) sulphate pentahydrate 97%
Iron(III) sulphate pentahydrate
CAS No.: 142906-29-4
Formule:
Fe₂H₁₀O₁₇S₃ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00149716 CAS nummer: 142906-29-4 UN: 3260 ADR: 8,III |
Specification Test Results
Appearance | Off-white to yellow to beige to grey Crystalline or granular powder |
Degree of hydration | May vary |
Titration with Na2S2O3 | 20.7 to 23 % (as Fe) |
Chloride (Cl) | ≤50 ppm |
Nitrate (NO3) | ≤200 ppm |
Phosphate (PO4) | ≤30 ppm |
Arsenic (As) | ≤5 ppm |
Copper (Cu) | ≤20 ppm |
Zinc (Zn) | ≤50 ppm |
Ferrous iron (Fe 2+) | ≤200 ppm |
Insoluble matter | ≤0.01 % (in HCl) |
Subst not pptd by NH3 | ≤0.05 % (as SO4) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...