Trimyristin 90%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
422090250
422090050
ACRO422090250EA
200
EUR
ACRO422090250
ACRO422090050
Trimyristin 90%
Trimyristin
CAS No.: 555-45-3
Formule:
C₄₅H₈₆O₆ Smeltpunt: 54…58 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00036229 CAS nummer: 555-45-3 |
Specification Test Results
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
TLC | ≥88 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...