N-Benzylidenemethylamine 99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
106010250
106011000
106010050
ACRO106010250EA
165
EUR
ACRO106010250
ACRO106011000
ACRO106010050
N-Benzylidenemethylamine 99%
N-Benzylidenemethylamine
CAS No.: 622-29-7
Formule:
C₈H₉N Kookpunt: 75…80 °C (18 mmHg) Dichtheid: 0,96 g/cm³ Vlampunt: >112 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008294 CAS nummer: 622-29-7 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5500 to 1.5540 (20°C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.002 % |
Titrable acid | ≤0.0003 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...