Perfluorodecahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture 90%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Perfluoro(decahydronaphthalene), Perfluorodecalin, Perfluorodecahydronaphthalene
130040250 130041000 130045000
ACRO130040250EA 48.5 EUR
ACRO130040250 ACRO130041000 ACRO130045000
Perfluorodecahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture 90%
Perfluorodecahydronaphthalene (mixture of cis and trans isomers)
Formule: C₁₀F₁₈
MW: 462.08
Kookpunt: 142 °C (760 mmHg)
Smeltpunt: –10 °C
Dichtheid: 1.9 g/cm³
Vlampunt: 59 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010626
CAS nummer: 306-94-5
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥88 %