Nicotinic acid N-oxide 99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
147781000
147780050
147785000
ACRO147781000EA
96.7
EUR
ACRO147781000
ACRO147780050
ACRO147785000
Nicotinic acid N-oxide 99%
Nicotinic acid N-oxide
CAS No.: 2398-81-4
Formule:
C₆H₅NO₃ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006201 CAS nummer: 2398-81-4 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | 98.5 to 101.5 % |
Specific optical rotation | -38.0° to -44.0° (20°C, 589 nm) (Neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...