Polypropyleenglycol, M.W. ∼ 425
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
192152500 192155000 192150025
ACRO192152500EA 76 EUR
ACRO192152500 ACRO192155000 ACRO192150025
Polypropyleenglycol, M.W. ∼ 425
Polypropyleenglycol
Formule: H(OCH₂CH₂CH₂)nOH
Kookpunt: 200 °C (760 mmHg)
Smeltpunt: –25 °C
Dichtheid: 1 g/cm³
Vlampunt: 185 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00084446
CAS nummer: 25322-69-4
Order Now
Specification Test Results
Appearance Clear viscous liquid
Infrared spectrum Conforms
Water ≤0.15 %
Color scale ≤70 APHA
Acid value ≤1 mg KOH/g
Hydroxyl value 250 to 276 mg KOH/g