Papaverine hydrochloride 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

207220050 207220250 207221000
ACRO207220050EA 58.9 EUR
ACRO207220050 ACRO207220250 ACRO207221000
Papaverine hydrochloride 99%
Papaverine hydrochloride

Appearance: White to beige Crystalline powder

Formule: C₂₀H₂₁NO₄·HCl
Smeltpunt: 226 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012745
CAS nummer: 61-25-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR