Phorbol-12-myristate-13-acetate 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: TPA, Phorbol 12-myristate 13-acetate, 12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, (1aR,1bS,4aR,7aS,7bS,8R,9R,9aS)-9a-Acetoxy-4a,7b-dihydroxy-3-(hydroxymethyl)-1,1,6,8-tetramethyl-5-oxo-1a,1b,4,4a,5,7a,7b,8,9,9a-decahydro-1H-cyclopropa[3,4]benzo[1,2-e]azulen-9-yl tetradecanoate
356150010 356150050
ACRO356150010EA 97.7 EUR
ACRO356150010 ACRO356150050
Phorbol-12-myristate-13-acetate 97%
Phorbol-12-myristate-13-acetate
CAS No.: 16561-29-8
Formule: C₃₆H₅₆O₈
MW: 616.836 g/mol
Smeltpunt: 72 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00036736
CAS nummer: 16561-29-8
Order Now
Specification Test Results
Appearance Colorless to white to light yellow Film or foam
HPLC ≥96.0 %