(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride 95%, 98% ee
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
362990010 362990050
ACRO362990010EA 191 EUR
ACRO362990010 ACRO362990050
(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride 95%, 98% ee
(R)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride
Appearance: White Powder
Formule: C₁₁H₁₆ClNO₂
MW: 229.704 g/mol
Smeltpunt: 119 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02259750
CAS nummer: 340188-50-3
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
HPLC ≥94 %
Enantiomeric excess ≥97.5 % (GC)