Tris(hydroxymethyl)phosphine 95%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
363840010 363840250
ACRO363840010EA 51 EUR
ACRO363840010 ACRO363840250
Tris(hydroxymethyl)phosphine 95%
Tris(hydroxymethyl)phosphine
CAS No.: 2767-80-8
Formule: C₃H₉O₃P
MW: 124.076 g/mol
Kookpunt: 111…113 °C (2.5 mmHg)
Smeltpunt: 48…56 °C
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00055382
CAS nummer: 2767-80-8
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to light yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
NMR ≥94 %