6,13-Pentacenedione 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

364270010 364270050
ACRO364270010EA 108 EUR
ACRO364270010 ACRO364270050
6,13-Pentacenedione 99%
6,13-Pentacenedione

CAS No.: 3029-32-1

Formule: C₂₂H₁₂O₂
Smeltpunt: 394 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003709
CAS nummer: 3029-32-1

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR