Chloro(1,5-cyclooctadiene)rhodium(I) dimer (>40,8% Rh)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (1,5-Cyclooctadiene)rhodium chloride dimer
211305000 211300050
ACRO211305000EA 543 EUR
ACRO211305000 ACRO211300050
Chloro(1,5-cyclooctadiene)rhodium(I) dimer (>40,8% Rh)
Chloro(1,5-cyclooctadiene)rhodium(I) dimer
Formule: C₁₆H₂₄Cl₂Rh₂
MW: 493.08 g/mol
Smeltpunt: 256 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012415
CAS nummer: 12092-47-6
Order Now
Specification Test Results
Appearance Orange to orange-brown Crystalline powder or crystals
Infrared spectrum Conforms
Rhodium (Rh) ≥40.8 %