Kaliumpolysulfiden, pure
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Potassium polysulphide, Liver of sulphur, Sulphurated potash
389592500
389590025
389590010
ACRO389592500EA
42.3
EUR
ACRO389592500
ACRO389590025
ACRO389590010
Kaliumpolysulfiden, pure
Kaliumpolysulfiden
Formule:
K₂(S)n Smeltpunt: 200…250 °C Dichtheid: 1,65 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00148074 CAS nummer: 37199-66-9 UN: 1382 ADR: 4.2,II |
Specification Test Results
Appearance | Ochre to brown Lumps |
Titration Iodometric | 42.0 to 44.0 % (K2S) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...