Kaliumhydroxide 1 N in ethanol
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Potash caustic, Caustic potash
393320010 393322500
ACRO393320010EA 42.9 EUR
ACRO393320010 ACRO393322500
Kaliumhydroxide 1 N in ethanol
Kaliumhydroxide
Formule: KOH
MW: 56.11 g/mol
Kookpunt: 78 °C (760 mmHg)
Smeltpunt: –117 °C
Dichtheid: 0.86 g/cm³
Vlampunt: 12 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003553
CAS nummer: 1310-58-3
UN: 2924
ADR: 3 II
Merck Index: 12,07806
Order Now
Specification Test Results
Appearance Clear colorless to yellow Solution
Strength 0.950 to 1.050 N