Dithiothreitol (DTT, Cleland's reagent) 1 M in water for biochemistry
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol
426380100 426380500 426382500
ACRO426380100EA 31.7 EUR
ACRO426380100 ACRO426380500 ACRO426382500
Dithiothreitol (DTT, Cleland's reagent) 1 M in water for biochemistry
Dithiothreitol (DTT, Cleland's reagent)
Formule: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004877
CAS nummer: 3483-12-3
UN: 3334
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Solution
Titration Iodimetric 0.90 to 1.10 M
UV at 500 nm A: ≤0.05