Benzylamine 99.5+%, AcroSeal®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: α-Aminotoluene
427231000 427238000
427231000.EA 70.8 EUR
427231000. 427238000.
Benzylamine 99.5+%, AcroSeal®
Benzylamine
Formule: (C₆H₅)CH₂NH₂
MW: 107.16 g/mol
Kookpunt: 185
Smeltpunt: –30 °C
Dichtheid: 0.98 g/cm³
Vlampunt: 72 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008106
CAS nummer: 100-46-9
UN: 2735
ADR: 8 II
Merck Index: 13,01127
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5424 to 1.5444 (20°C, 589 nm)
Water ≤0.3 %
GC ≥99.50 %