Water, special_quality TOC
Leverancier: EP SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Hydrogen oxide
112-01A/CTOC 111-04A/C-TOC
EPSC112-01A/CTOCEA 358 EUR
EPSC112-01A/CTOC EPSC111-04A/C-TOC
Water, special_quality TOC
Water
Highly purified low TOC water for sample and standards dilution, lab blank determination and equipment rinsing.
  • Packaged in pre-cleaned amber glass containers inside class 10 environment
  • Purity exceeds 17 MegaOhm resistivity
  • Low organic and inorganic impurities at time of packaging
  • Processed by reverse osmosis, activated carbon and ultraviolet TOC reduction
  • Filtered to 0,1 µ through hydrophobic membrane filters
  • Certified to contain <50 ppb TOC
1000 ml amber bottle is provided with PTFE/Silicone lined closure, Ø×H: 94×206 mm
4000 ml amber jug is provided with PP, low-shedding closure and PTFE/Silicone liner, Ø×H: 159×337 mm
Formule: H₂O
MW: 18.02 g/mol
Kookpunt: 100
Smeltpunt: 0
MDL Nummer: MFCD00011332
CAS nummer: 7732-18-5
Merck Index: 13,10098