Nucleic acid stains, PAGE GelRed™
Leverancier: Biotium
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41008-T
41008-500UL
41014
41008-TEA
42.8
EUR
41008-T
41008-500UL
41014.
Nucleic acid stains, PAGE GelRed™
Kleurstoffen
Kleuringen voor elektroforese
Stains specifically designed for DNA in polyacrylamide gels.
- More sensitive for agarose gel applications
- Two to three times more sensitive for dsDNA over RNA
- Impenetrable to latex or nitrile gloves and cell membranes
Unlike the highly mutagenic ethidium bromide (EtBr), PAGE GelRed are non-toxic and non-mutagenic in bacterial AMES tests.
PAGE GelRed can be imaged using a 254 nm UV transilluminator with an ethidium bromide filter.
Certificaten: CCR Title 22 for the management of hazardous waste.