Rotors for Sorvall™ RC 12 series
Leverancier: SORVALL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
77050 77080
SORV77050EA 21500 EUR
SORV77050 SORV77080
Rotors for Sorvall™ RC 12 series
Centrifuge Rotoren
Swinging-bucket rotors for use with the Thermo Scientific™ Sorvall™ RC12 series low-speed centrifuges.