Container, wide mouth, with hinged cap, Sterilin™
Leverancier: STERLIN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
100FLPLS 100FLS
SLIN100FLPLSEA 263 EUR
SLIN100FLPLS SLIN100FLS
Container, wide mouth, with hinged cap, Sterilin™
Monsternamepotjes Containers voor algemeen gebruik
Ideal for industrial sample collection, transport, storage and analysis.
  • Aseptically manufactured
  • Autoclavable at 121 °C with lid open
  • Frosted writing area for sample identification
  • Designed to stay vertical when opened
  • Available in natural and the food industry standard of blue
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904