Insole for women, ESD-PRO
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
204056/35 204056/36 204056/37 204056/38 204056/39 204056/40 204056/41 204056/42
114-7559EA 4.9 EUR
114-7559 114-7560 114-7561 114-7562 114-7563 114-7564 114-7565 114-7566
Insole for women, ESD-PRO
Inlegzolen voor schoenen
Antibacterial and anatomically shaped ESD insole.
  • Open cell structure of PU foam guarantees high moisture absorption
  • Activated carbon absorber reduces odour
  • Skin friendly materials improve the foot climate
For Elten Ladies shoes. Suitable for ESD safety shoes (electrical resistance of 105 to 107 Ohm).
Colour: Black