Cryogenic storage labels, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F134911301 F134913301 F134913302 F134916725 F134919501 F134921301 F134921302 F134923301 F134929501 F134929502
817-0416EA 365 EUR
817-0416 817-0411 817-0412 817-0413 817-0415 817-0419 817-0420 817-0414 817-0417 817-0418
Cryogenic storage labels, SP Bel-Art
Etiketten
The strong, permanent adhesive on these labels withstands cold storage temperatures and maintains adhesion to glass, plastic, cardboard and more, even in liquid nitrogen.
  • Organise and label samples for short or long-term cold storage
  • Ideal for environments down to −196 °C
  • Dot and rectangle shaped labels sized for 0,5 to 2,0 ml cryogenic tubes
  • Available in sheets or rolls