ESD safety shoes, lace-up, Officer
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
71307/47 71307/48 71307/40 71307/41 71307/42 71307/43 71307/44 71307/45 71307/46
ELTN71307/47EA 92.9 EUR
ELTN71307/47 ELTN71307/48 ELTN71307/40 ELTN71307/41 ELTN71307/42 ELTN71307/43 ELTN71307/44 ELTN71307/45 ELTN71307/46
ESD safety shoes, lace-up, Officer
Schoenen en laarzen
Business ESD safety shoes made from cowhide leather and a mono-density PU sole (Business).
  • Light leather lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (Business ESD)
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Black
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC