Biocompatible in-line filters, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A-429X A-431 A-430 A-428X
554-3038EA 195 EUR
554-3038 UPCHA-431 554-3039 UPCHA-428X
Biocompatible in-line filters, Upchurch Scientific®
Filters Lijnfilters
Inline filters consist of a stainless steel body and two PEEK end fittings.
  • Maximum recommended flow rate of 25 ml/min for A-430 and 10 ml/min for A-431
  • Designed to connect 1/16" OD or smaller tubing
  • Pressure rated to 6000 psi (414 bar)
  • 10-32 coned threads