In-line filters, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A-313 A-314 A-314-01 A-314-02
UPCHA-313EA 171 EUR
UPCHA-313 554-1781 554-3018 554-3019
In-line filters, Upchurch Scientific®
Filters Lijnfilters
Placed between pump and sample injection valve to trap particles released through normal piston seal wear.
  • For 1/16" OD tubing
  • Pressure rated to 6000 psi (414 bar)