Kaliumhexacyanoferraat (III) in ammonium hydroxide USP Test Solution (TS)
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide
85327.180EA 35.4 EUR
85327.180 85327.260
Kaliumhexacyanoferraat (III) in ammonium hydroxide USP Test Solution (TS)
Potassium hexacyanoferrate(III)
Ready to use test solution (TS).
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Formule: K₃Fe(CN)₆
MW: 329.25 g/mol
MDL Nummer: MFCD00011392
CAS nummer: 13746-66-2
EINECS: 0
Merck Index: 13,07712
Order Now
Specificaties
Environmentally Preferable