ESD Safety sandals, Velcro® fastening, Trainers, Scott Pro
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
726661/38 726661/39 726661/40 726661/41 726661/42 726661/43 726661/44 726661/45 726661/46 726661/47
ELTN726661/38EA 84.9 EUR
ELTN726661/38 ELTN726661/39 ELTN726661/40 ELTN726661/41 ELTN726661/42 ELTN726661/43 ELTN726661/44 ELTN726661/45 ELTN726661/46 ELTN726661/47
ESD Safety sandals, Velcro® fastening, Trainers, Scott Pro
Schoenen en laarzen
100% metal- and leather-free, lightweight ESD safety sandal made from microfibre and textile upper, a padded tongue plus a slip-resistant, dual-density TPU/PU sole (Trainers).
  • Climate-regulating, breathable textile lining
  • Full length fleece inlay sole (ESD Pro), black
  • TPU scuff cap protects the leather upper
  • Non metallic puncture resistant midsole
  • Composite toe protection cap (200 J)
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC