ESD safety shoes, lace-up, Dimension Pro, Ambition Boa Low
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
72752/36 72752/37 72752/38 72752/39 72752/40 72752/41 72752/42 72752/43 72752/44 72752/45 72752/46 72752/47 72752/48
114-7705EA 103.9 EUR
114-7705 114-7706 114-7707 114-7708 114-7709 114-7710 114-7711 114-7712 114-7713 114-7714 114-7715 114-7716 114-7717
ESD safety shoes, lace-up, Dimension Pro, Ambition Boa Low
Schoenen en laarzen
Anti-slip ESD safety shoes made from hydrophobised nubuck leather and hydrophobised textile upper, a full padded bellows tongue plus a slip resistant, dual-density TPU/PU sole (Dimension Pro).
  • Climate-regulating, breathable textile lining
  • Semi-orthopaedic, anatomically-shaped insole supports natural heel-to-toe foot movement
  • Replaceable insole; orthopaedic insoles possible
  • Sole prevents twisting and slipping
  • Lace- and buckle-free Boa® system can be opened and closed with just one hand
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Black/yellow
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC