ESD safety shoes, lace-up, Miss L10, Sensation Lady Low
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
74106/35 74106/36 74106/37 74106/38 74106/39 74106/40 74106/41 74106/42
114-7739EA 80.9 EUR
114-7739 114-7740 114-7741 114-7742 114-7743 114-7744 114-7745 114-7746
ESD safety shoes, lace-up, Miss L10, Sensation Lady Low
Schoenen en laarzen
Casual, vintage style ESD safety shoes for ladies made from hydrophobised material in 'denim look', and hydrophobised nubuck leather, a padded tongue plus a low profile, dual-density TPU/PU sole (Miss L10).
  • Bioactive fabric lining
  • Full length inlay sole (ESD Pro Lady), grey
  • Soft fleece ESD insole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected. The shoe is tailored to the ergonomics of female feet.
Colour: Black/grey
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC.