ESD safety ankle boots, lace-up, Miss L10, Vintage Lady Mid
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
74201/39 74201/35 74201/36 74201/37 74201/38 74201/40 74201/41 74201/42
114-8777EA 88.9 EUR
114-8777 114-8773 114-8774 114-8775 114-8776 114-8778 114-8779 114-8780
ESD safety ankle boots, lace-up, Miss L10, Vintage Lady Mid
Schoenen en laarzen
Casual, vintage style, environmental friendly manufactured ESD safety ankle boot made from waxy cowhide leather, a padded tongue plus a low profile, dual-density TPU/PU sole (Miss L10).
  • Bioactive fabric lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), grey
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects. The shoe is tailored to the ergonomics of female feet.
Colour: Grey
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.