Laboratory glass cleaner, deconex® 12 PA-x
Leverancier: Borer Chemie
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
516800.00-K05W 516800.00-K10W 516800.00-F10W
DECO516800.00-K05WEA 279 EUR
DECO516800.00-K05W DECO516800.00-K10W DECO516800.00-F10W
Laboratory glass cleaner, deconex® 12 PA-x
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Liquid, mildly alkaline cleaning concentrate for manual cleaning of laboratory glassware and reusable laboratory materials in ultrasonic and soaking baths.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • Perfect results even at low concentrations
  • Working solution suitable for multiple use and with a broad cleaning spectrum
  • High cleaning performance due to excellent swelling and dispersing capabilities
  • Effective against stubborn deposits
  • Especially recommended for sensitive materials
  • EDTA-, NTA- and chlorine-free
Order Now
Specificaties
Environmentally Preferable